Ethyl (Benzyloxy)acetate 97%

BLDPBD229166-5GEA 21 EUR
BLDPBD229166-5G BLDPBD229166-25G BLDPBD229166-1G BLDPBD229166-10G
Ethyl (Benzyloxy)acetate 97%
Ethyl (Benzyloxy)acetate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06204614
CAS-Nummer: 32122-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR