1-Hydroxy-2-naphthoesäure 98%
|
|
Synonyme:
1-Hydroxy-2-naphthalincarbonsäure
, 1-Hydroxynaphthalin-2-carbonsäure
BLDPBD22895-100GEA
15
EUR
BLDPBD22895-100G
BLDPBD22895-1000G
BLDPBD22895-500G
BLDPBD22895-25G
1-Hydroxy-2-naphthoesäure 98%
1-Hydroxy-2-naphthoesäure
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₁H₈O₃ Molecular Weight: 188,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003960 CAS-Nummer: 86-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...