Ethyl-7-bromimidazo[1,2-a]pyridin-2-carboxylat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD228288-100MGEA 21 EUR
BLDPBD228288-100MG BLDPBD228288-5G BLDPBD228288-25G BLDPBD228288-1G BLDPBD228288-250MG
Ethyl-7-bromimidazo[1,2-a]pyridin-2-carboxylat 98%
Ethyl-7-bromimidazo[1,2-a]pyridin-2-carboxylat

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₉BrN₂O₂
Molecular Weight: 269,10 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11505036
CAS-Nummer: 1187236-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR