2-Aminopyrazin 98%

Synonyme: AminopyrazinPyrazin-2-amin

BLDPBD2280-100GEA 54 EUR
BLDPBD2280-100G BLDPBD2280-25G BLDPBD2280-1000G BLDPBD2280-10G BLDPBD2280-500G
2-Aminopyrazin 98%
2-Aminopyrazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄H₅N₃
Molecular Weight: 95,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006137
CAS-Nummer: 5049-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR