3-Chlor-5-fluorsalicylaldehyd 97%

BLDPBD227860-1GEA 25 EUR
BLDPBD227860-1G BLDPBD227860-25G BLDPBD227860-5G
3-Chlor-5-fluorsalicylaldehyd 97%
3-Chlor-5-fluorsalicylaldehyd

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₄ClFO₂
Molecular Weight: 174,5569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039291
CAS-Nummer: 82128-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR