tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enylcarbamate 95%

BLDPBD226537-5GEA 260 EUR
BLDPBD226537-5G BLDPBD226537-100MG
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enylcarbamate 95%
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-3-enylcarbamate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₇H₃₀BNO₄
Molecular Weight: 323,235 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12032216
CAS-Nummer: 1251732-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR