2-(Pyrid-2-yloxy)benzaldehyde 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD225889-100MGEA 22 EUR
BLDPBD225889-100MG BLDPBD225889-1G BLDPBD225889-5G BLDPBD225889-250MG
2-(Pyrid-2-yloxy)benzaldehyde 95%
2-(Pyrid-2-yloxy)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₉NO₂
Molecular Weight: 199,205 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09025852
CAS-Nummer: 141580-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR