3-Chloro-5-nitrobenzyl bromide 97%

BLDPBD225583-1GEA 507 EUR
BLDPBD225583-1G BLDPBD225583-100MG BLDPBD225583-250MG
3-Chloro-5-nitrobenzyl bromide 97%
3-Chloro-5-nitrobenzyl bromide

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₅BrClNO₂
Molecular Weight: 250,477 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11505896
CAS-Nummer: 865624-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR