3'-Methyl-2,2,2-trifluoracetophenon 98%

BLDPBD224336-1GEA 31 EUR
BLDPBD224336-1G BLDPBD224336-5G BLDPBD224336-25G
3'-Methyl-2,2,2-trifluoracetophenon 98%
3'-Methyl-2,2,2-trifluoracetophenon

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇F₃O
Molecular Weight: 188,1465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461893
CAS-Nummer: 1736-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR