2'-Chloropropiophenone 95%

BLDPBD224318-250MGEA 37 EUR
BLDPBD224318-250MG BLDPBD224318-5G BLDPBD224318-1G BLDPBD224318-10G BLDPBD224318-25G
2'-Chloropropiophenone 95%
2'-Chloropropiophenone

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07787256
CAS-Nummer: 6323-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR