2-(2-Bromophenyl)pyridine 97%

BLDPBD223772-250MGEA 135 EUR
BLDPBD223772-250MG BLDPBD223772-100MG BLDPBD223772-5G BLDPBD223772-1G
2-(2-Bromophenyl)pyridine 97%
2-(2-Bromophenyl)pyridine

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₈BrN
Molecular Weight: 234,10 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15529755
CAS-Nummer: 109306-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR