(S)-α-(2-Oxiranyl)anisol 95%

BLDPBD22322-1GEA 25 EUR
BLDPBD22322-1G BLDPBD22322-25G BLDPBD22322-5G
(S)-α-(2-Oxiranyl)anisol 95%
(S)-α-(2-Oxiranyl)anisol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₀O₂
Molecular Weight: 150,1745 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06659026
CAS-Nummer: 71031-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR