[1,1'-Bis(diphenylphosphino)ferrocen]nickel(II)chlorid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD22123-25GEA 23 EUR
BLDPBD22123-25G BLDPBD22123-5G BLDPBD22123-10G BLDPBD22123-1G BLDPBD22123-100G
[1,1'-Bis(diphenylphosphino)ferrocen]nickel(II)chlorid 98%
[1,1'-Bis(diphenylphosphino)ferrocen]nickel(II)chlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₄H₂₈Cl₂FeNiP₂
Molecular Weight: 683,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270284
CAS-Nummer: 67292-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR