3-Amino-3-(4-bromphenyl)propionsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD220530-1GEA 15 EUR
BLDPBD220530-1G BLDPBD220530-5G BLDPBD220530-10G BLDPBD220530-250MG BLDPBD220530-100G BLDPBD220530-0.025 BLDPBD220530-25G
3-Amino-3-(4-bromphenyl)propionsäure 95%
3-Amino-3-(4-bromphenyl)propionsäure

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,0900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00187112
CAS-Nummer: 39773-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR