1-Benzyl-4-cyan-4-phenylpiperidin 98%

BLDPBD218895-1GEA 11 EUR
BLDPBD218895-1G BLDPBD218895-5G
1-Benzyl-4-cyan-4-phenylpiperidin 98%
1-Benzyl-4-cyan-4-phenylpiperidin
Formel: C₁₉H₂₀N₂
Molecular Weight: 276,38 g/mol
MDL: MFCD00044826
CAS-Nummer: 56243-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR