2-Brom-3-nitrobenzaldehyd 95%
|
|
BLDPBD218885-250MGEA
23
EUR
BLDPBD218885-250MG
BLDPBD218885-10G
BLDPBD218885-1G
BLDPBD218885-25G
BLDPBD218885-5G
2-Brom-3-nitrobenzaldehyd 95%
2-Bromo-3-nitrobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₄BrNO₃ Molecular Weight: 230,0200 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD15527595 CAS-Nummer: 90407-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...