5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride 97%

BLDPBD218771-5GEA 29 EUR
BLDPBD218771-5G BLDPBD218771-250MG BLDPBD218771-25G BLDPBD218771-100G BLDPBD218771-1G
5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride 97%
5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₀N₂O₂S·HCl
Molecular Weight: 234,70 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25542379
CAS-Nummer: 720720-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR