4-Chlor-(3-ethoxycarbonyl)phenylboronsäure 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD218066-5GEA 183 EUR
BLDPBD218066-5G BLDPBD218066-10G BLDPBD218066-1G BLDPBD218066-100MG BLDPBD218066-250MG
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure 96%
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₀BClO₄
Molecular Weight: 228,4400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08235062
CAS-Nummer: 874219-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR