3-Methoxy-5-methylphenylboronsäure 98%
|
|
BLDPBD217998-100MGEA
39
EUR
BLDPBD217998-100MG
BLDPBD217998-250MG
BLDPBD217998-5G
BLDPBD217998-1G
3-Methoxy-5-methylphenylboronsäure 98%
3-Methoxy-5-methylphenylboronsäure
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₈H₁₁BO₃ Molecular Weight: 165,9821 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09952026 CAS-Nummer: 725251-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...