3-Bromo-5-carboxybenzeneboronic acid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD217961-25GEA 246 EUR
BLDPBD217961-25G BLDPBD217961-1G BLDPBD217961-5G
3-Bromo-5-carboxybenzeneboronic acid 98%
3-Bromo-5-carboxybenzeneboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆BBrO₄
Molecular Weight: 244,835 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689487
CAS-Nummer: 913835-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR