2-Methoxy-1-naphthalinboronsäure 98%

BLDPBD217890-25GEA 220 EUR
BLDPBD217890-25G BLDPBD217890-250MG BLDPBD217890-5G BLDPBD217890-1G
2-Methoxy-1-naphthalinboronsäure 98%
2-Methoxy-1-naphthaleneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₁BO₃
Molecular Weight: 202,0100 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11044173
CAS-Nummer: 104116-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR