2-(Dimethylamino)-5-pyrimidinboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD217854-250MGEA 17 EUR
BLDPBD217854-250MG BLDPBD217854-100MG BLDPBD217854-5G BLDPBD217854-1G
2-(Dimethylamino)-5-pyrimidinboronsäure 98%
2-(Dimethylamino)-5-pyrimidineboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₆H₁₀BN₃O₂
Molecular Weight: 166,9735 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD10696933
CAS-Nummer: 756817-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR