6-Chloro-4-iodo-1H-indazole 95%
|
|
BLDPBD217054-1GEA
35
EUR
BLDPBD217054-1G
BLDPBD217054-250MG
BLDPBD217054-5G
6-Chloro-4-iodo-1H-indazole 95%
6-Chloro-4-iodo-1H-indazole
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₇H₄ClIN₂ Molecular Weight: 278,478 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07781383 CAS-Nummer: 885519-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...