4-Chlor-3-(trifluormethoxy)benzaldehyd 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD216284-250MGEA 18 EUR
BLDPBD216284-250MG BLDPBD216284-1G
4-Chlor-3-(trifluormethoxy)benzaldehyd 95%
4-Chloro-3-(trifluoromethoxy)benzaldehyde

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06660219
CAS-Nummer: 886499-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR