5-Brom-2-iodanisol 98%

BLDPBD213138-250MGEA 7 EUR
BLDPBD213138-250MG BLDPBD213138-100G BLDPBD213138-10G BLDPBD213138-25G BLDPBD213138-1G BLDPBD213138-5G
5-Brom-2-iodanisol 98%
5-Brom-2-iodanisol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₆BrIO
Molecular Weight: 312,9300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847482
CAS-Nummer: 791642-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR