4,6-Difluorsalicylaldehyd 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD212993-100MGEA 34 EUR
BLDPBD212993-100MG BLDPBD212993-250MG BLDPBD212993-5G BLDPBD212993-1G
4,6-Difluorsalicylaldehyd 97%
4,6-Difluorsalicylaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄F₂O₂
Molecular Weight: 158,1023 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03701048
CAS-Nummer: 136516-64-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR