4-(1-Naphthyl)-1-naphthalinboronsäure 98%

Synonyme: [1,1'-Binaphthalen]-4-ylboronic acid4-(Naphthalene-1-yl)-1-naphthalene boronic acid

BLDPBD212916-1GEA 51 EUR
BLDPBD212916-1G BLDPBD212916-250MG BLDPBD212916-5G
4-(1-Naphthyl)-1-naphthalinboronsäure 98%
4-(1-Naphthyl)-1-naphthaleneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂₀H₁₅BO₂
Lagertemperatur: Kühlschrank
MDL: MFCD16294426
CAS-Nummer: 363607-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR