tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD212544-250MGEA 47 EUR
BLDPBD212544-250MG BLDPBD212544-100MG BLDPBD212544-1G
tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate 95%
tert-Butyl 3-(iodomethyl)piperidine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₂₀INO₂
Molecular Weight: 325,186 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09953364
CAS-Nummer: 253177-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR