5-Formyl-2-methoxyphenyl Acetate 97%

BLDPBD212366-1GEA 17 EUR
BLDPBD212366-1G BLDPBD212366-25G BLDPBD212366-5G
5-Formyl-2-methoxyphenyl Acetate 97%
5-Formyl-2-methoxyphenyl Acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00266441
CAS-Nummer: 881-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR