D(-)-Penicillamin 98%

BLDPBD21229-1GEA 15 EUR
BLDPBD21229-1G BLDPBD21229-25G BLDPBD21229-5G BLDPBD21229-250MG BLDPBD21229-100G
D(-)-Penicillamin 98%
D(-)-Penicillamin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₁₁NO₂S
Molecular Weight: 149,2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00064302
CAS-Nummer: 52-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR