2-(Chloromethyl)-6-methylpyridine hydrochloride 98%

BLDPBD211277-1GEA 82 EUR
BLDPBD211277-1G BLDPBD211277-250MG
2-(Chloromethyl)-6-methylpyridine hydrochloride 98%
2-(Chloromethyl)-6-methylpyridine hydrochloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₈ClN·HCl
Molecular Weight: 178,06 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01691969
CAS-Nummer: 3099-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR