Methyl-5-fluor-2-iodbenzoat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD211045-5GEA 145 EUR
BLDPBD211045-5G BLDPBD211045-1G BLDPBD211045-100G BLDPBD211045-25G BLDPBD211045-10G BLDPBD211045-250MG
Methyl-5-fluor-2-iodbenzoat 97%
Methyl-5-fluor-2-iodbenzoat

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₆FIO₂
Molecular Weight: 280,0349 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12965043
CAS-Nummer: 1202897-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR