4'-Hydroxy-3'-methoxyacetophenon 98%
|
|
Synonyme:
Apocynin
, Acetovanillon
BLDPBD21023-5GEA
7
EUR
BLDPBD21023-5G
BLDPBD21023-500G
BLDPBD21023-0.025G
BLDPBD21023-100G
BLDPBD21023-25G
BLDPBD21023-1G
BLDPBD21023-10G
4'-Hydroxy-3'-methoxyacetophenon 98%
4'-Hydroxy-3'-methoxyacetophenon
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008747 CAS-Nummer: 498-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...