2-Chlornicotinoylchlorid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD21011-5GEA 22 EUR
BLDPBD21011-5G BLDPBD21011-25G
2-Chlornicotinoylchlorid 95%
2-Chlornicotinoylchlorid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₃Cl₂NO
Molecular Weight: 176,00 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051677
CAS-Nummer: 49609-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR