3-Amino-5-hydroxy-1H-pyrazol 95%
|
|
BLDPBD2097-10GEA
27
EUR
BLDPBD2097-10G
BLDPBD2097-5G
BLDPBD2097-25G
3-Amino-5-hydroxy-1H-pyrazol 95%
3-Amino-5-hydroxy-1H-pyrazole
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₃H₅N₃O Molecular Weight: 99,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233932 CAS-Nummer: 6126-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...