2-Indolboronsäurepinakolester 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD20944-25GEA 1814 EUR
BLDPBD20944-25G BLDPBD20944-10G BLDPBD20944-250MG BLDPBD20944-1G BLDPBD20944-100MG BLDPBD20944-5G
2-Indolboronsäurepinakolester 97%
2-Indolboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243,1092 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09751284
CAS-Nummer: 476004-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR