4-Carboxy-2-fluorphenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD208855-5GEA 143 EUR
BLDPBD208855-5G BLDPBD208855-1G BLDPBD208855-250MG BLDPBD208855-25G
4-Carboxy-2-fluorphenylboronsäure 97%
4-Carboxy-2-fluorphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆BFO₄
Molecular Weight: 183,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458473
CAS-Nummer: 851335-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR