Methyl-(4-bromphenyl)acetat 98%
|
|
BLDPBD20757-5GEA
7
EUR
BLDPBD20757-5G
BLDPBD20757-10G
BLDPBD20757-25G
BLDPBD20757-500G
BLDPBD20757-100G
Methyl-(4-bromphenyl)acetat 98%
Methyl-(4-bromphenyl)acetat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₉BrO₂ Molecular Weight: 229,0700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126844 CAS-Nummer: 41841-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...