3-Methoxy-5-(trifluormethyl)benzoesäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD206232-1GEA 17 EUR
BLDPBD206232-1G BLDPBD206232-250MG BLDPBD206232-5G
3-Methoxy-5-(trifluormethyl)benzoesäure 98%
3-Methoxy-5-(trifluoromethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025409
CAS-Nummer: 53985-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR