4-Fluoroisoquinoline 97%

BLDPBD20613-5GEA 278 EUR
BLDPBD20613-5G BLDPBD20613-25G BLDPBD20613-250MG BLDPBD20613-1G
4-Fluoroisoquinoline 97%
4-Fluoroisoquinoline

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₆FN
Molecular Weight: 147,15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06656128
CAS-Nummer: 394-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR