2-Chlor-5-fluornicotinsäure 97%
|
|
BLDPBD20568-1GEA
7
EUR
BLDPBD20568-1G
BLDPBD20568-100G
BLDPBD20568-25G
BLDPBD20568-5G
BLDPBD20568-10G
2-Chlor-5-fluornicotinsäure 97%
2-Chlor-5-fluornicotinsäure
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₃ClFNO₂ Molecular Weight: 175,5449 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092932 CAS-Nummer: 38186-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...