4-Methoxy-2-nitrobenzoic acid 98%

BLDPBD20301-5GEA 37 EUR
BLDPBD20301-5G BLDPBD20301-25G BLDPBD20301-1G BLDPBD20301-500G BLDPBD20301-100G BLDPBD20301-10G
4-Methoxy-2-nitrobenzoic acid 98%
4-Methoxy-2-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972109
CAS-Nummer: 33844-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR