2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid 97%

BLDPBD20222-25GEA 87 EUR
BLDPBD20222-25G BLDPBD20222-5G BLDPBD20222-250MG BLDPBD20222-100G BLDPBD20222-1G BLDPBD20222-10G
2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid 97%
2-Chlor-N-(2-ethyl-6-methylphenyl)acetamid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₄ClNO
Molecular Weight: 211,6880 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00267498
CAS-Nummer: 32428-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR