2-Brom-5-methoxy-4-methylanilin 98%

BLDPBD201956-5GEA 281 EUR
BLDPBD201956-5G BLDPBD201956-1G BLDPBD201956-250MG
2-Brom-5-methoxy-4-methylanilin 98%
2-Brom-5-methoxy-4-methylanilin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₁₀BrNO
Molecular Weight: 216,0751 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11841053
CAS-Nummer: 532440-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR