Methyl-4-tert-butylbenzoat 95%

BLDPBD19823-25GEA 7 EUR
BLDPBD19823-25G BLDPBD19823-500G BLDPBD19823-1G BLDPBD19823-10G BLDPBD19823-0.5G BLDPBD19823-1000G BLDPBD19823-100G
Methyl-4-tert-butylbenzoat 95%
Methyl-4-tert-butylbenzoat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008835
CAS-Nummer: 26537-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR