4-Fluor-3-methylbenzylamin 98%

BLDPBD19796-25GEA 98 EUR
BLDPBD19796-25G BLDPBD19796-1G BLDPBD19796-10G BLDPBD19796-5G
4-Fluor-3-methylbenzylamin 98%
4-Fluor-3-methylbenzylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631490
CAS-Nummer: 261951-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR