2-Brom-6-fluor-3-methylphenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD196767-5GEA 80 EUR
BLDPBD196767-5G BLDPBD196767-250MG BLDPBD196767-1G
2-Brom-6-fluor-3-methylphenylboronsäure 97%
2-Brom-6-fluor-3-methylphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇BBrFO₂
Molecular Weight: 232,8400 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12026712
CAS-Nummer: 957121-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR