2-Fluor-3-(trifluormethyl)phenylacetonitril 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD19579-25GEA 354 EUR
BLDPBD19579-25G BLDPBD19579-1G BLDPBD19579-5G
2-Fluor-3-(trifluormethyl)phenylacetonitril 98%
2-Fluor-3-(trifluormethyl)phenylacetonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅F₄N
Molecular Weight: 203,1363 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061178
CAS-Nummer: 239087-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR