4-Methoxyisophthalsäure 98%

BLDPBD19442-500GEA 474 EUR
BLDPBD19442-500G BLDPBD19442-100G BLDPBD19442-5G BLDPBD19442-25G
4-Methoxyisophthalsäure 98%
4-Methoxyisophthalsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈O₅
Molecular Weight: 196,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06208422
CAS-Nummer: 2206-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR