3-Amino-5-fluorpyridin 97%

BLDPBD19329-5GEA 18 EUR
BLDPBD19329-0.025G BLDPBD19329-5G BLDPBD19329-10G BLDPBD19329-100G BLDPBD19329-25G BLDPBD19329-1G
3-Amino-5-fluorpyridin 97%
3-Amino-5-fluoropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234073
CAS-Nummer: 210169-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR